Learn More
ATCC Staphylococcus aureus; 6 x 0.3mL
Supplier:  ATCC 6538PMINIPACK
Encompass
ATCC 6538P-MINI-PACK™ consists of 6 ready-to-use vials of ATCC 6538P™ frozen in 200 μL of glycerol stock, eliminating the need to rehydrate and culture the strain prior to use. Each vial is provided with a 2-D barcode for easy storage and tracking, as well as peel-off labels for fast and reliable recordkeeping.
Catalog No. 50-238-1562