Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phrixotoxin-1 TFA | 96.5% | 3548.30 | 1 MG
SDP

Catalog No. 5000399839
Encompass_Preferred

Phrixotoxin-1 TFA is a specific peptide inhibitor of the Kv4 potassium channel, derived from the venom of the theraphosid spider *Phrixotrichus auratus*. It is intended for research use only.

  • Purity of 96.5%
  • Molecular weight of 3548.30 (free base)
  • Appears as a solid, white to off-white powder
  • Solubility in H2O: ≥ 100 mg/mL
  • Powder storage at -80°C for 2 years or -20°C for 1 year
  • Storage in solvent at -80°C for 6 months or -20°C for 1 month
  • Functions as a potassium channel inhibitor

Catalog No. 50-003-99839 Supplier Medchemexpress LLC Supplier No. HYP2785A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phrixotoxin-1 TFA is a specific peptide inhibitor of the Kv4 potassium channel, derived from the venom of the theraphosid spider *Phrixotrichus auratus*. It is intended for research use only.

  • Purity of 96.5%
  • Molecular weight of 3548.30 (free base)
  • Appears as a solid, white to off-white powder
  • Solubility in H2O: ≥ 100 mg/mL
  • Powder storage at -80°C for 2 years or -20°C for 1 year
  • Storage in solvent at -80°C for 6 months or -20°C for 1 month
  • Functions as a potassium channel inhibitor

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.