Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ pHrodo™ Antibody Labeling Kits

Catalog No. P36020
Encompass_Preferred
Change view
Click to view available options
Quantity:
3 labelings kit
3 x 1 mg
3 x 100 μg
Excitation/Emission:
505/525 nm
560/585
560/585 nm
640/655 nm
Label or Dye:
pHrodo Deep Red
pHrodo Green
pHrodo Red
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Excitation/Emission Label or Dye
P36020 3 x 100 μg 560/585 nm pHrodo Red
P35355 3 x 100 μg 640/655 nm pHrodo Deep Red
P35356 3 x 1 mg 640/655 nm pHrodo Deep Red
P36021 3 x 1 mg 560/585 nm pHrodo Red
P36022 3 x 100 μg 505/525 nm pHrodo Green
P36023 3 x 1 mg 505/525 nm pHrodo Green
P36014 3 labelings kit 560/585 pHrodo Red
P36015 3 labelings kit 505/525 nm pHrodo Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. P36020 Supplier Invitrogen™ Supplier No. P36020
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Perform antibody internalization, endocytosis, and phagocytosis experiments with confidence and ease using the updated pH-sensitive, brightly fluorescing dyes and ready-to-use spin columns in the pHrodo Red and Green Antibody Labeling Kits.

Elevate biopharmaceutical research with pHrodo Antibody Labeling kits, designed to characterize antibody internalization, a critical step during antibody-drug conjugate research and development. Kits provide a simple, detailed protocol and advanced purification columns to easily label and purify antibodies and other large proteins. Amine-reactive pHrodo dyes covalently bind to antibodies and other proteins to produce bright, soluble conjugates whose fluorescence increases in acidic organelles including endosomes and lysosomes. Antibody internalization can be evaluated using flow cytometry, fluorescence microscopy, and high-content screening.

The pHrodo Red Antibody Labeling Kit and pHrodo Green Antibody Labeling Kit provide the reagents needed to easily label three purified protein or antibody samples with pHrodo Red or pHrodo Green dye. These kits contain amine-reactive, pH-sensitive pHrodo Red or Green STP ester dye that covalently binds to free lysines anywhere they occur in an antibody or protein.

The dyes included in this kit are improved versions of pHrodo Red and pHrodo Green dyes. They are optimized for the creation of bright and soluble bioconjugates that are used in the study of antibody internalization, endocytosis, phagocytosis, and for development of antibody-drug conjugates. These dyes are also more soluble than the original pHrodo Red and pHrodo Green dyes, making them useful for the labeling of antibodies that may otherwise precipitate out of solution during conjugation. pHrodo Red and pHrodo Green dyes are non-fluorescent outside the cell, but fluoresce bright red or bright green as the pH of their surroundings becomes more acidic, such as those found inside of phagosomes and endosomes.

The pHrodo Red and Green Microscale Labeling Kits offer the following benefits:
• Convenience—Labeling typically takes one hour, with less than 30 minutes of hands-on time
• Versatility—Labelling can be performed on 12-150 kDa molecular weight proteins
• Customization—Create your own probes for studying antibody internalization, phagocytosis, and endocytosis

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Flow Cytometry, Fluoresence Microscopy
Product Type Antibody Labeling Kit
Quantity 3 x 100 μg
Sufficient For 3 x 100 μg Reactions
Content And Storage Store refrigerated.
Detection Method Fluorescence
Excitation/Emission 560/585 nm
Labeling Scale 100 μg
Product Line pHrodo
Chemical Reactivity Amine
Reactive Moiety STP Ester
Solubility DMSO (Dimethylsulfoxide)
Label or Dye pHrodo Red
Spacer Mid-length, Pegylated
Color Red
Labeling Target Antibodies, Proteins
Flow Cytometer Laser Lines 561 nm
Label Type pHrodo
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.