Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Probes™ pHrodo™ Green Maleimide

Catalog No. P35370
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P35370 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P35370 Supplier Molecular Probes™ Supplier No. P35370
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

pHrodo™ Green Maleimide

The thiol-reactive pH-sensitive pHrodo™ Green Maleimide dye is suitable for the creation of bioconjugates to study endocytosis and phagocytosis. pHrodo™ Green dramatically increases fluorescence as the pH of its surroundings become more acidic.
  • Use pH-sensitive pHrodo™ Green Maleimide to make pH-sensitive bioconjugates of your choice
  • Get faster, more accurate results than with any other endocytosis or phagocytosis assay—no need for wash steps or quenchers
  • Multiplex with red fluorescent markers such as RFP, pHrodo™ Red, and many others

    The increase in fluorescence of pHrodo™ Green as pH changes from basic to acidic correlates with the acidification of intracellular vesicles, making it an ideal tool to study endocytosis or phagocytosis and their regulation by environmental factors, drugs, or pathogens. The spectral properties of pHrodo™ Green makes it useful for multi-color experiments. pHrodo™ Green can be detected using most standard green fluorescent filter sets and has been validated for use on a variety of platforms including flow cytometry, fluorescent microscopy, and high content screening (HCS). The lack of fluorescence of pHrodo™ Green in a typical extracellular environment eliminates the need for wash steps or quencher dyes in the experimental workflow.

    pHrodo™ Green Maleimide is a thiol-reactive dye that can be used to create pHrodo™ Green bioconjugates in aqueous buffer. The maleimide reacts with free sulphydryl groups produced by the reduction of cysteines in proteins or peptides. Maleimides are particularly useful for labeling antibodies as the dye will not attach to the antibody binding site. This reaction will result in a stable conjugate that can be used in live cell assays or stored for later use.

    pHrodo™ Green is also available in an amine-reactive form (see pHrodo™ Green STP), as well as a selection of ready-to-use conjugates (e.g., E. coli, S. aureus, and dextran). In addition, pHrodo™ Red reactive dyes and ready-to-use conjugates are available as a color alternative with the same properties.

    For Research Use Only. Not for human or animal therapeutic or diagnostic use.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Format Dried Down
    Quantity 1 mg
    Product Type Maleimide
    Solubility DMSO (Dimethylsulfoxide)
    Content And Storage 1 vial , store ≤ -20 °C, dessicate and protected from light.
    Chemical Reactivity Thiol
    Detection Method Fluorescence
    EC Extinction Coefficient 75000
    Reactive Group Thiol
    Label or Dye pHrodo™ Green
    Color Green
    Product Line pHrodo
    Emission 525
    Excitation 505
    Show More Show Less
    WARNING: Cancer - www.P65Warnings.ca.gov

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.