Learn More
Medchemexpress LLC HY-12063 10mg Medchemexpress, PHT-427 CAS:1191951-57-1 Purity:>98%
SDP
Catalog No. 502080947
Encompass_Preferred
Medchemexpress, HY-12063 10mg PHT-427 CAS:1191951-57-1 PHT-247 is an inhibitor of the pleckstrin homology (PH) domain of Akt, and it is also an inhibitor of PDPK1 with Kis of 2.7 µM and 5.2 µM and for Akt and PDPK1, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0947 Supplier Medchemexpress LLC Supplier No. HY1206310MG
Medchemexpress, HY-12063 10mg PHT-427 CAS:1191951-57-1 PHT-247 is an inhibitor of the pleckstrin homology (PH) domain of Akt, and it is also an inhibitor of PDPK1 with Kis of 2.7 µM and 5.2 µM and for Akt and PDPK1, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.