Learn More
Selleck Chemical LLC PHT-427 10mg 1191951-57-1 CS-0223
SDP
Catalog No. 508046
Encompass_Preferred
PHT-427 (CS-0223) is a dual Akt and PDPK1 inhibitor (high affinity binding for the PH domains of Akt and PDPK1) with Ki of 2.7 ?M and 5.2 ?M, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-804-6 Supplier Selleck Chemical LLC Supplier No. S155610MG
PHT-427 (CS-0223) is a dual Akt and PDPK1 inhibitor (high affinity binding for the PH domains of Akt and PDPK1) with Ki of 2.7 ?M and 5.2 ?M, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.