Learn More
Medchemexpress LLC PHT-7.3 | 1614225-93-2 | 99.29% | 100 MG
SDP
Catalog No. 5000342272
Encompass_Preferred
PHT-7.3 is a selective inhibitor of connector enhancer of kinase suppressor of Ras 1 (Cnk1) pleckstrin homology (PH) domain (Kd=4.7 μM). It inhibits mutant KRas, but not wild-type KRas cancer cell and tumor growth and signaling, demonstrating antitumor activity.
  • Selective inhibitor of Cnk1 PH domain.
  • Inhibits mutant KRas cancer cell and tumor growth.
  • Demonstrates antitumor activity.
  • Exhibits cytostatic antitumor activity in mut-KRas xenograft models.
Catalog No. 50-003-42272 Supplier Medchemexpress LLC Supplier No. HY128590100MG
PHT-7.3 is a selective inhibitor of connector enhancer of kinase suppressor of Ras 1 (Cnk1) pleckstrin homology (PH) domain (Kd=4.7 μM). It inhibits mutant KRas, but not wild-type KRas cancer cell and tumor growth and signaling, demonstrating antitumor activity.
  • Selective inhibitor of Cnk1 PH domain.
  • Inhibits mutant KRas cancer cell and tumor growth.
  • Demonstrates antitumor activity.
  • Exhibits cytostatic antitumor activity in mut-KRas xenograft models.