Learn More
Medchemexpress LLC PHT-7.3 | 1614225-93-2 | 99.3% | 10 MG
Supplier: Medchemexpress LLC HY12859010MG
PHT-7.3 is a selective inhibitor of the connector enhancer of kinase suppressor of Ras 1 (Cnk1) pleckstrin homology (PH) domain, with a Kd of 4.7 μM. It inhibits mutant KRas cancer cell and tumor growth and signaling, without affecting wild-type KRas. PHT-7.3 demonstrates antitumor activity and has shown cytostatic effects in mut-KRas(G12S) A549 and mut-KRasG12V H441 xenograft models at a dosage of 200 mg/kg.
- Selective inhibitor of Cnk1 PH-domain
- Inhibits mutant KRas cancer cell and tumor growth
- Demonstrates antitumor activity
- Shows cytostatic antitumor activity in mut-KRas xenograft models