Learn More
Medchemexpress LLC PHT-7.3 | 1614225-93-2 | 99.3% | 433.52 g·mol⁻¹ | C24H23N3O3S | 5 MG
SDP
Catalog No. 5000262118
Encompass_Preferred
PHT-7.3 is a research-grade small-molecule inhibitor that targets the pleckstrin homology (PH) domain of connector enhancer of kinase suppressor of Ras 1 (Cnk1) and shows activity against mutant KRAS signaling. Supplied as a high-purity solid for in vitro research use only.
  • Selective inhibitor of Cnk1 PH domain (Kd 4.7 μM).
  • Preferentially inhibits mutant KRAS signaling over wild-type KRAS.
  • High purity: 99.29%.
  • Soluble in DMSO at 62.5 mg/mL; may require ultrasonication.
  • Powder storage stability: -20°C for long-term, 4°C for short-term.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-62118 Supplier Medchemexpress LLC Supplier No. HY1285905MG
PHT-7.3 is a research-grade small-molecule inhibitor that targets the pleckstrin homology (PH) domain of connector enhancer of kinase suppressor of Ras 1 (Cnk1) and shows activity against mutant KRAS signaling. Supplied as a high-purity solid for in vitro research use only.
  • Selective inhibitor of Cnk1 PH domain (Kd 4.7 μM).
  • Preferentially inhibits mutant KRAS signaling over wild-type KRAS.
  • High purity: 99.29%.
  • Soluble in DMSO at 62.5 mg/mL; may require ultrasonication.
  • Powder storage stability: -20°C for long-term, 4°C for short-term.
  • Intended for research use only; not for human or clinical use.