Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical PHT PEG3 amidite | 25 g | CAS 2170381-91-4 | InChIKey UDTLXDXACDJZQQ-UHFFFAOYSA-N

Catalog No. 502364162
Encompass

PHT PEG3 amidite is a Amidite reagent (Subcategory: Amino Modifiers) sold by WuXi TIDES. Offered in 25 g. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: 2170381-91-4
- MDL: No data
- InChIKey: UDTLXDXACDJZQQ-UHFFFAOYSA-N
- Molecular Weight: 495.512761998
- Molecular Formula: C23H34N3O7P
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29280090
- Country Of Origin: China
- IUPAC: 2-cyanoethyl (2-(2-(2-((1,3-dioxoisoindolin-2-yl)oxy)ethoxy)ethoxy)ethyl) diisopropylphosphoramidite
- SMILES: CC(C)N(C(C)C)P(OCCOCCOCCON1C(C2=C(C=CC=C2)C1=O)=O)OCCC#N

Catalog No. 50-236-4162 Supplier STA Pharmaceutical Supplier No. STAAM1068625GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PHT PEG3 amidite is a Amidite reagent (Subcategory: Amino Modifiers) sold by WuXi TIDES. Offered in 25 g. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: 2170381-91-4
- MDL: No data
- InChIKey: UDTLXDXACDJZQQ-UHFFFAOYSA-N
- Molecular Weight: 495.512761998
- Molecular Formula: C23H34N3O7P
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29280090
- Country Of Origin: China
- IUPAC: 2-cyanoethyl (2-(2-(2-((1,3-dioxoisoindolin-2-yl)oxy)ethoxy)ethoxy)ethyl) diisopropylphosphoramidite
- SMILES: CC(C)N(C(C)C)P(OCCOCCOCCON1C(C2=C(C=CC=C2)C1=O)=O)OCCC#N

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.