Learn More
Cole-Parmer Oakton pHTestr 30 Waterproof Pocket Tester and Buffer Pack Bundle
Catalog No. 501961296 Shop All Cole-Parmer Products
Encompass_Preferred
The Oakton pHTestr 30 is great for outdoor applicationstake pH measurements in lakes and streams. Designed to be waterproof and float in water, you can even attach a line to the housing for easy retrieval. The replaceable sensor ensures easy maintenance, and the double-junction electrode handles wastewater, sulfides, heavy metals, and TRIS. The glass pH bulb is recessed to protect against breakage. The keypad clicks to confirm operation, and allows access to a variety of functions. Calibrate up to three pH points with auto-buffer recognition. Additional features include automatic temperature compensation (ATC), hold function, auto-off function, self-diagnostic error messages, and a three-segment battery-life indicator. The most common pH calibration buffers are also included!
Catalog No. 50-196-1296 Supplier Cole-Parmer Supplier No. 0675401
The Oakton pHTestr 30 is great for outdoor applicationstake pH measurements in lakes and streams. Designed to be waterproof and float in water, you can even attach a line to the housing for easy retrieval. The replaceable sensor ensures easy maintenance, and the double-junction electrode handles wastewater, sulfides, heavy metals, and TRIS. The glass pH bulb is recessed to protect against breakage. The keypad clicks to confirm operation, and allows access to a variety of functions. Calibrate up to three pH points with auto-buffer recognition. Additional features include automatic temperature compensation (ATC), hold function, auto-off function, self-diagnostic error messages, and a three-segment battery-life indicator. The most common pH calibration buffers are also included!