Learn More
Crescent Chemical Co Inc PHTHALALDEHYDE ANALYTICAL GRDE
SDP
Catalog No. 50248019
Encompass_Preferred
Phthalaldehyde analytical grade Especially purified for fluorimetric histidine determination (1). Reagent for amines and alkaloids (2) as well for amino acids (3, 4) and peptides (5, 6). Size - 5G Storage Conditions - +2 °C TO +8 °C Catalog Number
Catalog No. 50-248-019 Supplier Crescent Chemical Co Inc Supplier No. SE32800.01
Phthalaldehyde analytical grade Especially purified for fluorimetric histidine determination (1). Reagent for amines and alkaloids (2) as well for amino acids (3, 4) and peptides (5, 6). Size - 5G Storage Conditions - +2 °C TO +8 °C Catalog Number