Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phthalamic acid, 99%

Catalog No. AAL1243206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1243206 5 g
AAL1243214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1243206 Supplier Thermo Scientific Chemicals Supplier No. L1243206
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 88-97-1 | C8H7NO3 | 165.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-97-1
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.148
MDL Number MFCD00025476
InChI Key CYMRPDYINXWJFU-UHFFFAOYSA-N
Synonym phthalamic acid, phthalamidic acid, phthalamide acid, phthalic monoamide, phthalic acid monoamide, o-carbamoylbenzoic acid, benzoic acid, 2-aminocarbonyl, benzoic acid, o-carbamoyl, unii-v344h4pf3y, 2-aminocarbonyl benzoic acid
PubChem CID 6957
ChEBI CHEBI:50736
IUPAC Name 2-carbamoylbenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)N)C(=O)O

Specifications

Melting Point 140°C to 143°C
Quantity 5 g
Beilstein 1868205
Formula Weight 165.15
Percent Purity 99%
Chemical Name or Material Phthalamic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-871-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.