Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phthalanilide 98.0+%, TCI America™
SDP

Catalog No. P028525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P028525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P028525G Supplier TCI America Supplier No. P028525G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16497-41-9
Molecular Formula C20H16N2O2
Molecular Weight (g/mol) 316.36
MDL Number MFCD00059671
InChI Key ZYACJWRVLBPZNG-UHFFFAOYSA-N
Synonym N,N′C-Diphenylphthalamide, Phthaldianilide
PubChem CID 85459
IUPAC Name 1-N,2-N-diphenylbenzene-1,2-dicarboxamide
SMILES C1=CC=C(C=C1)NC(=O)C2=CC=CC=C2C(=O)NC3=CC=CC=C3

Specifications

Melting Point 225°C
Quantity 25 g
Formula Weight 316.36
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Phthalanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.