Learn More
Hach Company Phthalate-Phosphate Reagent Powder Pillows, 10 mL, pk/100
Catalog No. 501996353 Shop All Hach Company Products
Encompass
For low-level determination of nickel and cobalt by the PAN method. Hach method 8078. Sample Size: 10-mL Pack of 100 Powder Pillows. Replacement reagent for Cobalt/Nickel Reagent Set - 2651600.
Catalog No. 50-199-6353 Supplier Hach Company Supplier No. 2615199
For low-level determination of nickel and cobalt by the PAN method. Hach method 8078. Sample Size: 10-mL Pack of 100 Powder Pillows. Replacement reagent for Cobalt/Nickel Reagent Set - 2651600.