Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phthalates in Polyvinyl Chloride Standard, 8 Components, SPEX CertiPrep™
SDP

Catalog No. CRMPVC001 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1.5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CRMPVC001 1.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CRMPVC001 Supplier SPEX CertiPrep Supplier No. CRMPVC001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Phthalates in Polyvinyl Chloride
Name Note Designed to meet the regulations required for testing of PVC toys and other consumer products
Packaging 2mL Amber Prescored Glass Ampule
Methods For Consumer Product Safety Commission Method CPSC-CH-C1001-09.1
Solvent or Matrix Polyvinyl Chloride
Quantity 1.5 g
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.