Learn More
Medchemexpress LLC Phthalazine | 253-52-1 | 130.15 | 1 ML
SDP
Catalog No. 5000447688
Encompass_Preferred
Phthalazine is a substrate for human aldehyde oxidase 1, which can lead to the production of reactive oxygen species (ROS) and subsequent enzyme inactivation. It is a nitrogen-containing organic heterocyclic compound. Phthalazine has demonstrated antitumor activity in various cancers including blood cancer, breast cancer, colon cancer, lung cancer, and renal cancer.
  • Substrate for human aldehyde oxidase 1
  • Leads to the production of reactive oxygen species (ROS)
  • Nitrogen-containing organic heterocyclic compound
  • Demonstrated antitumor activity in various cancers
Catalog No. 50-004-47688 Supplier Medchemexpress LLC Supplier No. HYW00201610MM/1ML
Phthalazine is a substrate for human aldehyde oxidase 1, which can lead to the production of reactive oxygen species (ROS) and subsequent enzyme inactivation. It is a nitrogen-containing organic heterocyclic compound. Phthalazine has demonstrated antitumor activity in various cancers including blood cancer, breast cancer, colon cancer, lung cancer, and renal cancer.
  • Substrate for human aldehyde oxidase 1
  • Leads to the production of reactive oxygen species (ROS)
  • Nitrogen-containing organic heterocyclic compound
  • Demonstrated antitumor activity in various cancers