Learn More
Medchemexpress LLC Phthalazine | 253-52-1 | 130.15 | 100 G
SDP
Catalog No. 5000447477
Encompass_Preferred
Phthalazine is a nitrogen-containing organic heterocyclic compound and a substrate for human aldehyde oxidase 1, which can lead to the production of reactive oxygen species (ROS) and subsequent enzyme inactivation. It also exhibits antitumor activity.
  • Nitrogen-containing organic heterocyclic compound
  • Substrate for human aldehyde oxidase 1
  • Leads to the production of reactive oxygen species (ROS) and subsequent enzyme inactivation
  • Exhibits antitumor activity in various cancers
Catalog No. 50-004-47477 Supplier Medchemexpress LLC Supplier No. HYW002016100G
Phthalazine is a nitrogen-containing organic heterocyclic compound and a substrate for human aldehyde oxidase 1, which can lead to the production of reactive oxygen species (ROS) and subsequent enzyme inactivation. It also exhibits antitumor activity.
  • Nitrogen-containing organic heterocyclic compound
  • Substrate for human aldehyde oxidase 1
  • Leads to the production of reactive oxygen species (ROS) and subsequent enzyme inactivation
  • Exhibits antitumor activity in various cancers