Learn More
Medchemexpress LLC Phthalazine | 253-52-1 | 130.15 | 5 G
SDP
Catalog No. 5000448833
Encompass_Preferred
This organic heterocyclic compound serves as a substrate for human aldehyde oxidase 1, potentially generating reactive oxygen species and leading to enzyme inactivation. It has shown promising activity against various cancers, including blood, breast, colon, lung, and renal cancers.
  • Substrate for human aldehyde oxidase 1.
  • Nitrogen-containing organic heterocyclic compound.
  • Exhibits antitumor activity.
Catalog No. 50-004-48833 Supplier Medchemexpress LLC Supplier No. HYW0020165G
This organic heterocyclic compound serves as a substrate for human aldehyde oxidase 1, potentially generating reactive oxygen species and leading to enzyme inactivation. It has shown promising activity against various cancers, including blood, breast, colon, lung, and renal cancers.
  • Substrate for human aldehyde oxidase 1.
  • Nitrogen-containing organic heterocyclic compound.
  • Exhibits antitumor activity.