Learn More
Medchemexpress LLC Phthalazine | 253-52-1 | 130.15 | 50 G
SDP
Catalog No. 5000447337
Encompass_Preferred
Phthalazine is a nitrogen-containing organic heterocyclic compound that acts as a substrate for human aldehyde oxidase 1. This interaction can lead to the production of reactive oxygen species (ROS) and subsequent enzyme inactivation. It has demonstrated antitumor activity in various cancers.
  • Substrate for human aldehyde oxidase 1
  • Involved in reactive oxygen species (ROS) production and enzyme inactivation
  • Exhibits antitumor activity in blood cancer, breast cancer, colon cancer, lung cancer, and renal cancer
  • Nitrogen-containing organic heterocyclic compound
Catalog No. 50-004-47337 Supplier Medchemexpress LLC Supplier No. HYW00201650G
Phthalazine is a nitrogen-containing organic heterocyclic compound that acts as a substrate for human aldehyde oxidase 1. This interaction can lead to the production of reactive oxygen species (ROS) and subsequent enzyme inactivation. It has demonstrated antitumor activity in various cancers.
  • Substrate for human aldehyde oxidase 1
  • Involved in reactive oxygen species (ROS) production and enzyme inactivation
  • Exhibits antitumor activity in blood cancer, breast cancer, colon cancer, lung cancer, and renal cancer
  • Nitrogen-containing organic heterocyclic compound