Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phthalazine 98.0+%, TCI America™
SDP

Catalog No. P069425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P069425G 25 g
P06945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P069425G Supplier TCI America Supplier No. P069425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 253-52-1
Molecular Formula C8H6N2
Molecular Weight (g/mol) 130.15
MDL Number MFCD00006908
InChI Key LFSXCDWNBUNEEM-UHFFFAOYSA-N
Synonym 2,3-benzodiazine, benzo d pyridazine, 2,3-diazanaphthalene, beta-phenodiazine, .beta.-phenodiazine, unii-91y28dm24n, ccris 6894, phthalazin, benzo-orthodiazine
PubChem CID 9207
ChEBI CHEBI:36597
IUPAC Name phthalazine
SMILES C1=CC=C2C=NN=CC2=C1

Specifications

Melting Point 93°C
Color White-Yellow
Boiling Point 317°C
Quantity 25 g
Formula Weight 130.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Phthalazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.