Learn More
Cayman Chemical Phthalazinone pyrazole
Supplier: Cayman Chemical 107355 MG
A potent inhibitor of Aurora A kinase (IC50 = 31 nM); does not inhibit Aurora B kinase at doses up to 100 μM; inhibits the proliferation of HCT116, COLO 205, and MCF-7 cells (IC50 = 7.8, 2.9, and 1.6 μM, respectively)