Learn More
Cayman Chemical Phthalazinone pyrazole, Purity: ≥98%, CAS: 880487-62-7, 1 mg
SDP
Catalog No. 502888039
Encompass_Preferred
A potent inhibitor of Aurora A kinase (IC50 = 31 nM); does not inhibit Aurora B kinase at doses up to 100 μM; inhibits the proliferation of HCT116, COLO 205, and MCF-7 cells (IC50 = 7.8, 2.9, and 1.6 μM, respectively)
Catalog No. 50-288-8039 Supplier Cayman Chemical Supplier No. 107351 MG
A potent inhibitor of Aurora A kinase (IC50 = 31 nM); does not inhibit Aurora B kinase at doses up to 100 μM; inhibits the proliferation of HCT116, COLO 205, and MCF-7 cells (IC50 = 7.8, 2.9, and 1.6 μM, respectively)