Learn More
Medchemexpress LLC Phthalazinone pyrazole | 880487-62-7 | MFCD28399120 | 98.0% | 317.34 | C18H15N5O | 10MG
SDP
Catalog No. 5000235209
Encompass_Preferred
Phthalazinone pyrazole is a potent, selective, orally active small-molecule inhibitor of Aurora-A kinase used for research in cell biology and oncology. It has a biochemical IC50 of 0.031 μM against Aurora-A and shows cellular activity in multiple cancer cell lines.
  • Potent Aurora-A kinase inhibition (IC50 0.031 μM).
  • Demonstrated cellular activity in COLO 205, HCT-116, and MCF7 cell lines.
  • High purity suitable for research (98.0%).
  • Well-characterized physicochemical properties (C18H15N5O; MW 317.34).
  • Available as solid and solution formats for assay flexibility.
  • Supplied with a datasheet containing analytical and handling information.
Catalog No. 50-002-35209 Supplier Medchemexpress LLC Supplier No. HY1256410MG
Phthalazinone pyrazole is a potent, selective, orally active small-molecule inhibitor of Aurora-A kinase used for research in cell biology and oncology. It has a biochemical IC50 of 0.031 μM against Aurora-A and shows cellular activity in multiple cancer cell lines.
  • Potent Aurora-A kinase inhibition (IC50 0.031 μM).
  • Demonstrated cellular activity in COLO 205, HCT-116, and MCF7 cell lines.
  • High purity suitable for research (98.0%).
  • Well-characterized physicochemical properties (C18H15N5O; MW 317.34).
  • Available as solid and solution formats for assay flexibility.
  • Supplied with a datasheet containing analytical and handling information.