Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phthalazinone pyrazole | 880487-62-7 | MFCD28399120 | 98.0% | 317.34 | C18H15N5O | 10MM 1ML
SDP

Catalog No. 5000237864
Encompass_Preferred

Phthalazinone pyrazole is a potent, selective Aurora-A kinase inhibitor (IC50 = 0.031 μM) reported to arrest mitosis, induce apoptosis in proliferating cells, and suppress epithelial-mesenchymal transition during hepatocyte-like cell differentiation. It is supplied as a DMSO solution or as solid powder for research use.

  • Potent, selective Aurora-A kinase inhibitor (IC50 = 0.031 μM).
  • Reported to arrest mitosis and induce apoptosis in proliferating cells.
  • Suppresses epithelial-mesenchymal transition during cell differentiation.
  • Available as 10 mM in DMSO (1 mL) and multiple solid sizes.
  • Molecular formula C18H15N5O; molecular weight 317.34.
  • Purity 98.0%; store solid at 4°C protected from light; in solution store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-37864 Supplier Medchemexpress LLC Supplier No. HY1256410MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Phthalazinone pyrazole is a potent, selective Aurora-A kinase inhibitor (IC50 = 0.031 μM) reported to arrest mitosis, induce apoptosis in proliferating cells, and suppress epithelial-mesenchymal transition during hepatocyte-like cell differentiation. It is supplied as a DMSO solution or as solid powder for research use.

  • Potent, selective Aurora-A kinase inhibitor (IC50 = 0.031 μM).
  • Reported to arrest mitosis and induce apoptosis in proliferating cells.
  • Suppresses epithelial-mesenchymal transition during cell differentiation.
  • Available as 10 mM in DMSO (1 mL) and multiple solid sizes.
  • Molecular formula C18H15N5O; molecular weight 317.34.
  • Purity 98.0%; store solid at 4°C protected from light; in solution store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.