Learn More
Medchemexpress LLC Phthalazinone pyrazole | 880487-62-7 | MFCD28399120 | 98.0% | 317.34 | C18H15N5O | 25MG
SDP
Catalog No. 5000237405
Encompass_Preferred
Phthalazinone pyrazole is a small-molecule Aurora-A kinase inhibitor provided as a brown to gray solid for biochemical and cellular research. The compound has formula C18H15N5O, molecular weight 317.34 g·mol⁻1, and a reported purity of 98.0%. It is offered in multiple solid sizes and as 10 mM solutions in DMSO, and should be stored protected from light at 4°C (in solvent: -80°C for long term storage).
  • Potent Aurora-A kinase inhibitor with low-nanomolar activity.
  • High reported purity for assay reliability.
  • Multiple solid pack sizes and solution formats available.
  • Brown to gray solid; handle and store protected from light.
  • Accompanied by supporting documents such as data sheet, COA, and SDS.
Catalog No. 50-002-37405 Supplier Medchemexpress LLC Supplier No. HY1256425MG
Phthalazinone pyrazole is a small-molecule Aurora-A kinase inhibitor provided as a brown to gray solid for biochemical and cellular research. The compound has formula C18H15N5O, molecular weight 317.34 g·mol⁻1, and a reported purity of 98.0%. It is offered in multiple solid sizes and as 10 mM solutions in DMSO, and should be stored protected from light at 4°C (in solvent: -80°C for long term storage).
  • Potent Aurora-A kinase inhibitor with low-nanomolar activity.
  • High reported purity for assay reliability.
  • Multiple solid pack sizes and solution formats available.
  • Brown to gray solid; handle and store protected from light.
  • Accompanied by supporting documents such as data sheet, COA, and SDS.