Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phthalazinone pyrazole | 880487-62-7 | MFCD28399120 | 98.0% | 317.34 | C18H15N5O | 5MG
SDP

Catalog No. 5000237135
Encompass_Preferred

Phthalazinone pyrazole is a small-molecule Aurora-A kinase inhibitor used in research on cell cycle regulation, apoptosis, and mitosis. It is supplied as a solid and as a DMSO solution for biochemical and cellular assays.

  • Potent Aurora-A kinase inhibitor for research use.
  • Molecular formula C18H15N5O, molecular weight 317.34 g·mol⁻¹.
  • Purity ≥98.0% by HPLC.
  • Available as solid (5 mg-100 mg) and solution (10 mM in DMSO, 1 mL).
  • Store at 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).
  • Provided with datasheet, SDS, and COA for characterization.

Catalog No. 50-002-37135 Supplier Medchemexpress LLC Supplier No. HY125645MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phthalazinone pyrazole is a small-molecule Aurora-A kinase inhibitor used in research on cell cycle regulation, apoptosis, and mitosis. It is supplied as a solid and as a DMSO solution for biochemical and cellular assays.

  • Potent Aurora-A kinase inhibitor for research use.
  • Molecular formula C18H15N5O, molecular weight 317.34 g·mol⁻¹.
  • Purity ≥98.0% by HPLC.
  • Available as solid (5 mg-100 mg) and solution (10 mM in DMSO, 1 mL).
  • Store at 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).
  • Provided with datasheet, SDS, and COA for characterization.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.