Learn More
Sigma Aldrich Fine Chemicals Biosciences Phthaldialdehyde suitable for HPLC fluorimetric detection of amino acids, >=99% (HPLC), powder | 643-79-8 | MFCD00003335 | 250MG
Catalog No. 501853807
Encompass
Phthaldialdehyde suitable for HPLC fluorimetric detection of amino acids, >=99% (HPLC), powder | Purity: >=99% (HPLC) | Mol Wt: 134.13 | 643-79-8 | MFCD00003335 | 250MG
Catalog No. 50-185-3807 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P0657250MG
Phthaldialdehyde suitable for HPLC fluorimetric detection of amino acids, >=99% (HPLC), powder | Purity: >=99% (HPLC) | Mol Wt: 134.13 | 643-79-8 | MFCD00003335 | 250MG