Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phthalic acid Bis(3,5,5-tri-methylhexyl) ester, 99.6%, MP Biomedicals™

Catalog No. ICN15624891 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15624891 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15624891 Supplier MP Biomedicals, Inc Supplier No. 0215624891

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 28553-12-0
Molecular Formula C26H42O4
Molecular Weight (g/mol) 418.618
InChI Key HBGGXOJOCNVPFY-UHFFFAOYSA-N
Synonym diisononyl phthalate, 1,2-benzenedicarboxylic acid, diisononyl ester, bis 7-methyloctyl benzene-1,2-dicarboxylate, bis 7-methyloctyl phthalate, dinp branched, phthalic acid bis 7-methyloctyl ester, dsstox_cid_2521, dsstox_rid_78653, dsstox_gsid_28665, 1,2-benzenedicarboxylic acid, bis 7-methyloctyl ester
PubChem CID 590836
ChEBI CHEBI:35459
IUPAC Name bis(7-methyloctyl) benzene-1,2-dicarboxylate
SMILES CC(C)CCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCC(C)C

Specifications

Color Colorless
Density 0.973 - 0.977g/cm3 at 20°C (Palatinol N)
Quantity 1 kg
Formula Weight 418.6
Percent Purity 0.996
Physical Form Liquid
Chemical Name or Material Phthalic acid Bis(3, 5, 5-tri-methylhexyl) ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.