Learn More
Medchemexpress LLC Phthalic acid mono-2-ethylhexyl ester | 4376-20-9 | MFCD00041500 | 97.7% | 278.34 g·mol⁻¹ | C16H22O4 | 50 MG
SDP
Catalog No. 5000301655
Encompass_Preferred
Mono-(2-ethylhexyl) phthalate (MEHP) is the primary bioactive metabolite of di(2-ethylhexyl) phthalate and is provided as an analytical standard for biochemical, toxicology, and environmental studies. It is offered as solid packs and as ready-to-use DMSO solutions, with analytical-grade purity suitable for method development, calibration, and in vitro assays.
  • High purity of 97.7% for analytical applications.
  • Molecular formula C16H22O4 and molecular weight 278.34 g·mol⁻¹.
  • Supplied as small solid packs and DMSO solutions for convenience.
  • Suitable for LC-MS and GC-MS method development and calibration.
  • Appropriate for in vitro pharmacology and toxicology studies.
Catalog No. 50-003-01655 Supplier Medchemexpress LLC Supplier No. HYW01839250MG
Mono-(2-ethylhexyl) phthalate (MEHP) is the primary bioactive metabolite of di(2-ethylhexyl) phthalate and is provided as an analytical standard for biochemical, toxicology, and environmental studies. It is offered as solid packs and as ready-to-use DMSO solutions, with analytical-grade purity suitable for method development, calibration, and in vitro assays.
  • High purity of 97.7% for analytical applications.
  • Molecular formula C16H22O4 and molecular weight 278.34 g·mol⁻¹.
  • Supplied as small solid packs and DMSO solutions for convenience.
  • Suitable for LC-MS and GC-MS method development and calibration.
  • Appropriate for in vitro pharmacology and toxicology studies.