Learn More
Sigma Aldrich Fine Chemicals Biosciences Phthalic anhydride Reagent
Catalog No. 502897424
Encompass
Phthalic anhydride is an industrially important chemical. It can be prepared from o-xylene or naphthalene. It is widely used for the industrial preparation of plasticizers for PVC (polyvinyl chloride).Phthalic anhydride can be synthesized via oxidation of o-xylene in the presence of V2O5/TiO2 monolithic catalyst supported by aluminium honeycomb.
Catalog No. 50-289-7424 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 12573316KG
Phthalic anhydride is an industrially important chemical. It can be prepared from o-xylene or naphthalene. It is widely used for the industrial preparation of plasticizers for PVC (polyvinyl chloride).Phthalic anhydride can be synthesized via oxidation of o-xylene in the presence of V2O5/TiO2 monolithic catalyst supported by aluminium honeycomb.