Learn More
Medchemexpress LLC Phthalide | 87-41-2 | MFCD00005906 | 99.9% | 134.13 g/mol | C8H6O2 | 50 G
SDP
Catalog No. 5000300065
Encompass_Preferred
Phthalide (CAS 87-41-2) is a small-molecule organic compound (C8H6O2, MW 134.13 g/mol) used as a chemical scaffold and reagent in organic synthesis and biochemical research. It is a solid powder commonly used to prepare phthalide derivatives with reported anti-inflammatory, antimicrobial, and antioxidant activities. Recommended storage: powder -20°C (3 years) or 4°C (2 years).
  • Chemical scaffold for synthesis of derivatives.
  • Suitable for biochemical assay reagent preparation.
  • Reported biological activities: anti-inflammatory, antimicrobial, antioxidant.
  • Stable solid with defined storage conditions.
  • Available in research pack sizes for laboratory use.
Catalog No. 50-003-00065 Supplier Medchemexpress LLC Supplier No. HYW01582050G
Add to Cart
Add to Cart
Phthalide (CAS 87-41-2) is a small-molecule organic compound (C8H6O2, MW 134.13 g/mol) used as a chemical scaffold and reagent in organic synthesis and biochemical research. It is a solid powder commonly used to prepare phthalide derivatives with reported anti-inflammatory, antimicrobial, and antioxidant activities. Recommended storage: powder -20°C (3 years) or 4°C (2 years).
  • Chemical scaffold for synthesis of derivatives.
  • Suitable for biochemical assay reagent preparation.
  • Reported biological activities: anti-inflammatory, antimicrobial, antioxidant.
  • Stable solid with defined storage conditions.
  • Available in research pack sizes for laboratory use.