Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phthalide, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass Bottle
Plastic bottle
87-41-2
C8H6O2
134.13
MFCD00005906
WNZQDUSMALZDQF-UHFFFAOYSA-N
phthalide,isobenzofuran-1 3h-one,1 3h-isobenzofuranone,1-phthalanone,phthalolactone,2-benzofuran-1 3h-one,isobenzofuranone,phthalanone,3h-isobenzofuran-1-one,2-hydroxymethylbenzoic acid, gamma-lactone
6885
CHEBI:38085
3H-2-benzofuran-1-one
C1C2=CC=CC=C2C(=O)O1
87-41-2
C8H6O2
134.13
MFCD00005906
WNZQDUSMALZDQF-UHFFFAOYSA-N
phthalide,isobenzofuran-1 3h-one,1 3h-isobenzofuranone,1-phthalanone,phthalolactone,2-benzofuran-1 3h-one,isobenzofuranone,phthalanone,3h-isobenzofuran-1-one,2-hydroxymethylbenzoic acid, gamma-lactone
6885
CHEBI:38085
3H-2-benzofuran-1-one
C1C2=CC=CC=C2C(=O)O1
Boiling Point 290°C
Flash Point 152°C
Packaging Glass Bottle
Melting Point 70°C to 73°C
Color White to Beige
Quantity 5g
Infrared Spectrum Authentic
Assay Percent Range 99%
Beilstein 17,31
Fieser 05,537; 08,402
Solubility Information Solubility in water: sparingly soluble
Formula Weight 134.13
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material Phthalide, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 201-744-0

RTECSNumber : TI3520000

TSCA : TSCA