Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phthalimide, 98%, Spectrum™ Chemical
SDP

Catalog No. 18612368 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-368 1 kg Poly Pail
18-612-369 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-612-368 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P11411KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 85-41-6
Molecular Formula C8H5NO2
Molecular Weight (g/mol) 147.13
InChI Key XKJCHHZQLQNZHY-UHFFFAOYSA-N
IUPAC Name 2,3-dihydro-1H-isoindole-1,3-dione
SMILES O=C1NC(=O)C2=CC=CC=C12

Specifications

CAS Max % 1
Melting Point 231° to 236°C
Packaging Poly Pail
Quantity 1 kg
Percent Purity 0.98
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.