Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phthalimide, Potassium salt, 99%, MP Biomedicals™

Catalog No. ICN10264980 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10264980 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10264980 Supplier MP Biomedicals, Inc Supplier No. 0210264980

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1074-82-4
Molecular Formula C8H4KNO2
Molecular Weight (g/mol) 185.22
MDL Number MFCD00005887
InChI Key FYRHIOVKTDQVFC-UHFFFAOYSA-M
Synonym potassium phthalimide, potassium 1,3-dioxoisoindolin-2-ide, n-potassiophthalimide, phthalimide potassium salt, n-potassium phthalimide, unii-x6kka27dil, phthalimide, potassium salt, potassium phthalamide, 1h-isoindole-1,3 2h-dione, potassium salt, x6kka27dil
PubChem CID 3356745
IUPAC Name potassium 1,3-dioxo-2,3-dihydro-1H-isoindol-2-ide
SMILES [K+].O=C1[N-]C(=O)C2=CC=CC=C12

Specifications

Color Yellow
Assay Percent Range 99%
Quantity 100 g
Formula Weight 185.2
Physical Form Powder
Chemical Name or Material Phthalimide, Potassium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.