Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phthalocyanine | 574-93-6 | 98.0% | 10 G
SDP

Catalog No. 5000232940
Encompass_Preferred

Phthalocyanine is a dark purple to black macrocyclic organic dye and photosensitizer supplied as a solid for laboratory research, with properties suitable for photodynamic and materials studies.

  • Solid, dark purple to black appearance.
  • Molecular formula C32H18N8; molecular weight 514.54 g/mol.
  • Purity 98.0% as specified by the SDS.
  • Suitable for photodynamic and materials research applications.
  • Available as a 10 G pack size for laboratory use.

Catalog No. 50-002-32940 Supplier Medchemexpress LLC Supplier No. HY12131010G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phthalocyanine is a dark purple to black macrocyclic organic dye and photosensitizer supplied as a solid for laboratory research, with properties suitable for photodynamic and materials studies.

  • Solid, dark purple to black appearance.
  • Molecular formula C32H18N8; molecular weight 514.54 g/mol.
  • Purity 98.0% as specified by the SDS.
  • Suitable for photodynamic and materials research applications.
  • Available as a 10 G pack size for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.