Learn More
Medchemexpress LLC Phthalocyanine | 574-93-6 | 98.0% | 5 G
SDP
Catalog No. 5000233741
Encompass_Preferred
Phthalocyanine is a macrocyclic organic compound used as a photosensitizer in photodynamic research and sensor development. It exhibits light-activated antimicrobial activity and can be conjugated for targeted therapeutic and diagnostic applications.
  • Photosensitizer with activity against bacterial biofilms.
  • Applicable to photodynamic therapy and antimicrobial studies.
  • Compatible with conjugation chemistry for targeted delivery or detection.
  • Solid, dark purple to black powder, organosoluble.
  • Available in small laboratory quantities for research use.
Catalog No. 50-002-33741 Supplier Medchemexpress LLC Supplier No. HY1213105G
Phthalocyanine is a macrocyclic organic compound used as a photosensitizer in photodynamic research and sensor development. It exhibits light-activated antimicrobial activity and can be conjugated for targeted therapeutic and diagnostic applications.
  • Photosensitizer with activity against bacterial biofilms.
  • Applicable to photodynamic therapy and antimicrobial studies.
  • Compatible with conjugation chemistry for targeted delivery or detection.
  • Solid, dark purple to black powder, organosoluble.
  • Available in small laboratory quantities for research use.