Learn More
Strem, An Ascensus Company CAS# 574-93-6. 5g. Phthalocyanine. MFCD00005085
SDP
Catalog No. 5090109062
Encompass
CAS# 574-93-6. 5g. Phthalocyanine. MFCD00005085. Molecular Weight: 514.55. Molecular Formula: C32H18N8. Color/form: blue black pwdr. Strem# 07-1700. http://www.strem.com/catalog/v/07-1700/
Catalog No. 50-901-09062 Supplier Strem, An Ascensus Company Supplier No. 0717005G
CAS# 574-93-6. 5g. Phthalocyanine. MFCD00005085. Molecular Weight: 514.55. Molecular Formula: C32H18N8. Color/form: blue black pwdr. Strem# 07-1700. http://www.strem.com/catalog/v/07-1700/