Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phthalocyanine, MP Biomedicals

Catalog No. ICN216700 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN216700 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN216700 Supplier MP Biomedicals, Inc Supplier No. 0521670005
Add to Cart
Add to Cart

Chemical Identifiers

CAS 574-93-6
Molecular Formula C32H18N8
Molecular Weight (g/mol) 514.552
InChI Key IEQIEDJGQAUEQZ-UHFFFAOYSA-N
Synonym phthalocyanine, pigment blue 16, 29h,31h-phthalocyanine, phthalocyanin, heliogen blue g, polymon blue g, lionol blue kw, irgazin blue 3gt, ftalocianina, ci pigment blue 16
PubChem CID 5282330
ChEBI CHEBI:34921
SMILES C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C(N4)N=C6C7=CC=CC=C7C(=N6)N=C8C9=CC=CC=C9C(=N8)N=C2N3

Specifications

Quantity 10 g
Formula Weight 514.55
Chemical Name or Material Phthalocyanine
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.