Learn More
New England Biolabs, Inc. Phusion GC Buffer Pack – 6 ml (4 x 1.5 ml)
SDP
Catalog No. 50995081
Encompass
New England Biolabs supplies a unique 5X reaction buffer for high GC targets and supplemental reagents for use with Thermo Scientific Phusion High-Fidelity DNA Polymerase and associated products, all of which are manufactured and quality controlled at New England Biolabs.
Catalog No. 50-995-081 Supplier New England Biolabs, Inc. Supplier No. B0519S
New England Biolabs supplies a unique 5X reaction buffer for high GC targets and supplemental reagents for use with Thermo Scientific Phusion High-Fidelity DNA Polymerase and associated products, all of which are manufactured and quality controlled at New England Biolabs.