Learn More
New England Biolabs, Inc. Phusion™ High-Fidelity DNA Polymerase – 500 units
SDP
Supplier:  New England Biolabs, Inc. M0530L
Encompass
High-Fidelity DNA Polymerases are important for applications in which the DNA sequence needs to be correct after amplification. Its unique structure, a novel Pyrococcus-like enzyme fused with a processivity-enhancing domain, increases fidelity and speed. Phusion DNA Polymerase is an ideal choice for cloning and can be used for long or difficult amplicons. With an error rate > 50-fold lower than that of Taq DNA Polymerase and 6-fold lower than that of Pyrococcus furiosus DNA Polymerase, Phusion is one of the most accurate thermostable polymerases available. Phusion DNA Polymerase possesses 5' to 3' polymerase activity, 3' to 5' exonuclease activity and will generate blunt-ended products. Phusion DNA Polymerase is supplied with standard 5X Phusion HF Buffer, as well as 5X Phusion GC Buffer, which can be used for complex or GC-rich templates. Each of these buffers contains MgCl2 (1.5 mM at the final [1X] reaction concentration).
Catalog No. 5001234