Learn More
Med Vet International Betadine Solution, 1 Gallon
Catalog No. 502828829
Encompass_Preferred
Betadine Solution is a topical, water-based antiseptic microbicide used to kill bacteria, fungi and viruses. Contains 5% povidone-iodine (0.5% available iodine). Use full strength for: Preoperative prepping of skin and mucous membranes. Preventing bacterial infection. Emergency antisepsis of minor lacerations, abrasions, and burns. Postoperative application to surgical incisions. Avian | canine | cattle | equine | feline | goat | miscellaneous | poultry | sheep | swine
Catalog No. 50-282-8829 Supplier Med Vet International Supplier No. PHVBETASOL1G
Betadine Solution is a topical, water-based antiseptic microbicide used to kill bacteria, fungi and viruses. Contains 5% povidone-iodine (0.5% available iodine). Use full strength for: Preoperative prepping of skin and mucous membranes. Preventing bacterial infection. Emergency antisepsis of minor lacerations, abrasions, and burns. Postoperative application to surgical incisions. Avian | canine | cattle | equine | feline | goat | miscellaneous | poultry | sheep | swine