Learn More
Medchemexpress LLC Phycocyanins, C- | 11016-15-2 | 10 MG
SDP
Catalog No. 5000297261
Encompass_Preferred
C-phycocyanin (C-PC) is a water-soluble blue phycobiliprotein pigment derived from cyanobacteria, used in research as a fluorescent label and as a nutritional supplement.
  • Water-soluble blue protein pigment.
  • Broad research uses: fluorescent labeling, biochemical assays, and supplements.
  • Absorption maximum near 615-620 nm (lambda max).
  • Typically supplied as a lyophilized powder; store at 4°C and protect from light.
  • Available in multiple small-scale sizes for laboratory use.
Catalog No. 50-002-97261 Supplier Medchemexpress LLC Supplier No. HYD102510MG
C-phycocyanin (C-PC) is a water-soluble blue phycobiliprotein pigment derived from cyanobacteria, used in research as a fluorescent label and as a nutritional supplement.
  • Water-soluble blue protein pigment.
  • Broad research uses: fluorescent labeling, biochemical assays, and supplements.
  • Absorption maximum near 615-620 nm (lambda max).
  • Typically supplied as a lyophilized powder; store at 4°C and protect from light.
  • Available in multiple small-scale sizes for laboratory use.