Learn More
Selleck Chemical LLC Phycocyanobilin-5mg
SDP
Catalog No. 502389742
Encompass_Preferred
Phycocyanobilin (PCB) is an open-chain tetrapyrrole chromophore covalently bonded to both polypeptide chains of C-Phycocyanin (C-PC), the most represented biliprotein of Spirulina platensis, the mechanisms by which it protected cells included the reduction of oxidative stress damage, which could contribute to its clinical efficacy for the treatment of neurodegenerative diseases.This product has poor solubility, animal experiments are available, cell experiments please choose carefully!
Catalog No. 50-238-9742 Supplier Selleck Chemical LLC Supplier No. E26935MG
Phycocyanobilin (PCB) is an open-chain tetrapyrrole chromophore covalently bonded to both polypeptide chains of C-Phycocyanin (C-PC), the most represented biliprotein of Spirulina platensis, the mechanisms by which it protected cells included the reduction of oxidative stress damage, which could contribute to its clinical efficacy for the treatment of neurodegenerative diseases.This product has poor solubility, animal experiments are available, cell experiments please choose carefully!