Learn More
Aviva Systems Biology Corp PHYH Antibody - N-terminal region (ARP56681_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56681_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PHYH. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PHYH. Alternate names: LN1, LNAP1, PAHX, PHYH1, RDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-193