Learn More
Aviva Systems Biology Corp PHYHIP Antibody - N-terminal region (ARP33878_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33878_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PHYHIP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PHYHIP. Alternate names: KIAA0273, PAHX-AP, PAHXAP1, DYRK1AP3ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55184