Learn More
Medchemexpress LLC Phyllanthin | 10351-88-9 | 1 MG
SDP
Catalog No. 5000419015
Encompass_Preferred
Phyllanthin is a potent anticancer agent exhibiting diverse biological activities. It effectively inhibits MOLT-4 cell viability, induces apoptosis, and hinders cell migration and invasion. Beyond its anticancer properties, Phyllanthin also demonstrates anti-fibrotic effects through the down-regulation of the TGF signaling pathway, specifically targeting ALK5 and Smad2/3. Additionally, it possesses notable anti-inflammatory and antibacterial characteristics, making it a versatile compound for various research applications.
  • Inhibits MOLT-4 cell viability
  • Increases apoptosis
  • Inhibits cell migration and invasion
  • Exerts anti-fibrotic effects by down-regulating the TGF signaling pathway via ALK5 and Smad2/3
  • Possesses anti-inflammatory properties
  • Exhibits antibacterial characteristics
Catalog No. 50-004-19015 Supplier Medchemexpress LLC Supplier No. HYN41071MG
Phyllanthin is a potent anticancer agent exhibiting diverse biological activities. It effectively inhibits MOLT-4 cell viability, induces apoptosis, and hinders cell migration and invasion. Beyond its anticancer properties, Phyllanthin also demonstrates anti-fibrotic effects through the down-regulation of the TGF signaling pathway, specifically targeting ALK5 and Smad2/3. Additionally, it possesses notable anti-inflammatory and antibacterial characteristics, making it a versatile compound for various research applications.
  • Inhibits MOLT-4 cell viability
  • Increases apoptosis
  • Inhibits cell migration and invasion
  • Exerts anti-fibrotic effects by down-regulating the TGF signaling pathway via ALK5 and Smad2/3
  • Possesses anti-inflammatory properties
  • Exhibits antibacterial characteristics