Learn More
Medchemexpress LLC Phyllanthin | 10351-88-9 | 5 MG
SDP
Catalog No. 5000419749
Encompass_Preferred
Phyllanthin is an effective oral anticancer agent. It inhibits MOLT-4 cell viability, increases apoptosis, and restricts cell migration and invasion. This compound also exhibits anti-fibrotic effects by down-regulating the TGF signaling pathway. Furthermore, Phyllanthin possesses anti-inflammatory and antibacterial properties, and is predicted to have strong activity against COVID-19.
  • Inhibits MOLT-4 cell viability and increases apoptosis
  • Restricts cell migration and invasion
  • Exerts anti-fibrotic effects via TGF signaling pathway downregulation
  • Possesses anti-inflammatory and antibacterial properties
  • Predicted to have strong activity against COVID-19
  • Inhibits TGF-β1
  • Inhibits LPS-induced proinflammatory responses in U937 macrophages
Catalog No. 50-004-19749 Supplier Medchemexpress LLC Supplier No. HYN41075MG
Phyllanthin is an effective oral anticancer agent. It inhibits MOLT-4 cell viability, increases apoptosis, and restricts cell migration and invasion. This compound also exhibits anti-fibrotic effects by down-regulating the TGF signaling pathway. Furthermore, Phyllanthin possesses anti-inflammatory and antibacterial properties, and is predicted to have strong activity against COVID-19.
  • Inhibits MOLT-4 cell viability and increases apoptosis
  • Restricts cell migration and invasion
  • Exerts anti-fibrotic effects via TGF signaling pathway downregulation
  • Possesses anti-inflammatory and antibacterial properties
  • Predicted to have strong activity against COVID-19
  • Inhibits TGF-β1
  • Inhibits LPS-induced proinflammatory responses in U937 macrophages