Learn More
Medchemexpress LLC Phylloquinone | 84-80-0 | 450.70 | 500 MG
SDP
Catalog No. 5000416740
Encompass_Preferred
Vitamin K1 (Standard) is the analytical standard of Vitamin K1. This product is intended for research and analytical applications. Vitamin K1 a naturally occurring vitamin required for blood coagulation and bone and vascular metabolism.
  • This product is intended for research and analytical applications.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • It is commonly used in qualitative, quantitative and methodological research experiments in high-performance liquid chromatography, gas chromatography and mass spectrometry.
Catalog No. 50-004-16740 Supplier Medchemexpress LLC Supplier No. HYN0684R500MG
Vitamin K1 (Standard) is the analytical standard of Vitamin K1. This product is intended for research and analytical applications. Vitamin K1 a naturally occurring vitamin required for blood coagulation and bone and vascular metabolism.
  • This product is intended for research and analytical applications.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • It is commonly used in qualitative, quantitative and methodological research experiments in high-performance liquid chromatography, gas chromatography and mass spectrometry.