Learn More
Cayman Chemical Phyllanthin, Purity: ≥98%, CAS: 10351-88-9, 10 mg
SDP
Catalog No. 502951840
Encompass_Preferred
A bioactive lignan component of P. amarus with antioxidative and hepatoprotective properties; scavenges DPPH radicals (IC50 = 7.4 µM) and alleviates carbon tetrachloride-induced lipid peroxidation and cytotoxicity in a human hepatoma HepG2 cell line
Catalog No. 50-295-1840 Supplier Cayman Chemical Supplier No. 1174510 MG
A bioactive lignan component of P. amarus with antioxidative and hepatoprotective properties; scavenges DPPH radicals (IC50 = 7.4 µM) and alleviates carbon tetrachloride-induced lipid peroxidation and cytotoxicity in a human hepatoma HepG2 cell line